EMAIL THIS PAGE TO A FRIEND
Trials
Study design and rationale of 'Influence of Cilostazol-based triple anti-platelet therapy on ischemic complication after drug-eluting stent implantation (CILON-T)' study: A multicenter randomized trial evaluating the efficacy of Cilostazol on ischemic vascular complications after drug-eluting stent implantation for coronary heart disease.
PMID 20735821
Abstract
Current guidelines recommend dual anti-platelet therapy, aspirin and clopidogrel, for patients treated with drug-eluting stent for coronary heart disease. In a few small trials, addition of cilostazol on dual anti-platelet therapy (triple anti-platelet therapy) showed better late luminal loss. In the real-world unselected patients with coronary heart disease, however, the effect of cilostazol on platelet reactivity and ischemic vascular events after drug-eluting stent implantation has not been tested. It is also controversial whether there is a significant interaction between lipophilic statin and clopidogrel. CILON-T trial was a prospective, randomized, open-label, multi-center, near-all-comer trial to demonstrate the superiority of triple anti-platelet therapy to dual anti-platelet therapy in reducing 6 months' major adverse cardiovascular/cerebrovascular events, composite of cardiac death, nonfatal myocardial infarction, target lesion revascularization and ischemic stroke. It also tested whether triple anti-platelet therapy is superior to dual anti-platelet therapy in inhibiting platelet reactivity in patients receiving percutaneous coronary intervention with drug-eluting stent. Total 960 patients were randomized to receive either dual anti-platelet therapy or triple anti-platelet therapy for 6 months and also, randomly stratified to either lipophilic statin (atorvastatin) or non-lipophilic statin (rosuvastatin) indefinitely. Secondary endpoints included all components of major adverse cardiovascular/cerebrovascular events, platelet reactivity as assessed by VerifyNow P2Y12 assay, effect of statin on major adverse cardiovascular/cerebrovascular events, bleeding complications, and albumin-to-creatinine ratio to test the nephroprotective effect of cilostazol. Major adverse cardiovascular/cerebrovascular events will also be checked at 1, 2, and 3 years to test the 'legacy' effect of triple anti-platelet therapy that was prescribed for only 6 months after percutaneous coronary intervention. CILON-T trial will give powerful insight into whether triple anti-platelet therapy is superior to dual anti-platelet therapy in reducing ischemic events and platelet reactivity in the real-world unselected patients treated with drug-eluting stent for coronary heart disease. Also, it will verify the laboratory and clinical significance of drug interaction between lipophilic statin and clopidogrel. National Institutes of Health Clinical Trials Registry (ClinicalTrials.gov identifier# NCT00776828).