EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
A mecA-negative strain of methicillin-resistant Staphylococcus aureus with high-level β-lactam resistance contains mutations in three genes.
PMID 20805396
Abstract
We previously generated a ceftobiprole-resistant Staphylococcus aureus strain after high inoculum serial passage of a mecA-negative variant of strain COL (R. Banerjee, M. Gretes, L. Basuino, N. Strynadka, and H. F. Chambers, Antimicrob. Agents Chemother. 52:2089-2096, 2008). Genome resequencing of this strain, CRB, revealed that it differs from its parent by five single-nucleotide polymorphisms in three genes, specifically, those encoding PBP4, a low-molecular-weight penicillin-binding protein, GdpP, a predicted signaling protein, and AcrB, a cation multidrug efflux transporter. CRB displayed resistance to a variety of β-lactams but was hypersusceptible to cefoxitin.