EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and antifungal activity of benzofuran-5-ols.
PMID 20851600
Abstract
Benzofuran-5-ol derivatives were synthesized and tested for in vitro antifungal activity against Candida, Aspergillus species, and Cryptococcus neoformans. Among them tested, many benzofuran-5-ols showed good antifungal activity. The results suggest that benzofuran-5-ols would be promising antifungal agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
39579
2,5-Dimethyl-1,4-benzoquinone, ≥98.0%
C8H8O2