EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Novel apramycin resistance gene apmA in bovine and porcine methicillin-resistant Staphylococcus aureus ST398 isolates.
PMID 20876371
Abstract
A novel apramycin resistance gene, apmA, was detected on the ca.-40-kb resistance plasmid pAFS11 from bovine methicillin-resistant Staphylococcus aureus (MRSA) of sequence type 398 (ST398). The apmA gene coded for a protein of 274 amino acids that was related only distantly to acetyltransferases involved in chloramphenicol or streptogramin A resistance. NsiI deletion of apmA resulted in a 16- to 32-fold decrease in the apramycin MICs. An apmA-specific PCR identified this gene in one additional bovine and four porcine MRSA ST398 isolates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2024
Apramycin sulfate salt, ≥95% (TLC)
C21H41N5O11 · xH2SO4