EMAIL THIS PAGE TO A FRIEND
Onkologie
Mitomycin C in patients with gynecological malignancies.
PMID 20926904
Abstract
Mitomycin C (MMC) is an effective cytostatic agent used in the treatment of patients with gynecological malignancies and breast carcinoma. This review presents and discusses the current treatment options with MMC in patients with breast, cervical, and vulvar carcinomas, as well as rarer gynecological malignancies. New combinations and developments are also presented and their potential clinical relevance is examined. Consequently, also for the next years a MMC-containing chemotherapy continues to be a relevant part of an individualized therapy despite numerous innovative new drugs, especially for the salvage therapy of metastatic breast cancer and the simultaneous radiochemotherapy of other gynecological malignancies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000378 Mitomycin, European Pharmacopoeia (EP) Reference Standard
C15H18N4O5