EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Solvent-free aromatic C-H functionalisation/halogenation reactions.
PMID 20931130
Abstract
The solvent-free, palladium-catalysed reaction of anilides with CuCl(2) in the presence or absence of copper acetate yields ortho-chlorinated anilides in good to excellent yields, even on a large scale (100 mmol). By contrast, the equivalent reactions with copper bromide, either solvent free or in 1,2-dichloroethane, in the presence or absence of palladium, under air or inert conditions, gave the products of simple electrophilic bromination. Mechanistic studies highlighted the involvement of palladacyclic intermediates, one of which was characterised crystallographically, which undergo subsequent reaction with copper(II) chloride to yield the chlorinated anilide products.