EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and antihyperlipidemic activity of novel coumarin bisindole derivatives.
PMID 20932744
Abstract
A series of novel coumarin bisindole heterocycles were synthesized following an uncommon method and evaluated for their antihyperlipidemic activity in hyperlipidemic hamster model. Among 12 compounds tested, the compound 5e showed potent antihyperlipidemic activity and was found to decrease the plasma triglyceride levels (TG) by 55%, total cholesterol (TC) by 20%, accompanied by an increase in HDL-C/TC ratio by 42% in hyperlipidemic rats to a greater degree than some of the reference statins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D165603
1,2-Dimethylindole, 99%
C10H11N
193984
1-Methylindole, ≥97%
C9H9N