EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Cell physiology
Cathelicidin LL-37 peptide regulates endothelial cell stiffness and endothelial barrier permeability.
PMID 20943960
Abstract
LL-37 peptide is a multifunctional host defense molecule essential for normal immune responses to infection or tissue injury. In this study we assess the impact of LL-37 on endothelial stiffness and barrier permeability. Fluorescence microscopy reveals membrane localization of LL-37 after its incubation with human umbilical vein endothelial cells (HUVECs). A concentration-dependent increase in stiffness was observed in HUVECs, bovine aortic endothelial cells (BAECs), human pulmonary microvascular endothelial cells, and mouse aorta upon LL-37 (0.5-5 μM) addition. Stiffening of BAECs by LL-37 was blocked by P2X7 receptor antagonists and by the intracellular Ca²(+) chelator BAPTA-AM. Increased cellular stiffness correlated with a decrease in permeability of HUVEC cell monolayers after LL-37 addition compared with nontreated cells, which was similar to the effect observed upon treatment with sphingosine 1-phosphate, and both treatments increased F-actin content in the cortical region of the cells. These results suggest that the antiinflammatory effect of LL-37 at the site of infection or injury involves an LL-37-mediated increase in cell stiffening that prevents increased pericellular permeability. Such a mechanism may help to maintain tissue fluid homeostasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1076
BAPTA-AM, ≥95% (HPLC)
C34H40N2O18
S9666
Sphingosine 1-phosphate, ≥95%, powder
C18H38NO5P