EMAIL THIS PAGE TO A FRIEND
Cell division
JAB1/CSN5: a new player in cell cycle control and cancer.
PMID 20955608
Abstract
c-Jun activation domain-binding protein-1 (Jab1) acts as a modulator of intracellular signaling and affects cellular proliferation and apoptosis, through its existence as a monomer or as the fifth component of the constitutive photomorphogenic-9 signalosome (CSN5). Jab1/CSN5 is involved in transcription factor specificity, deneddylation of NEDD8, and nuclear-to-cytoplasmic shuttling of key molecules. Jab1/CSN5 activities positively and negatively affect a number of pathways, including integrin signaling, cell cycle control, and apoptosis. Also, more recent studies have demonstrated the intriguing roles of Jab1/CSN5 in regulating genomic instability and DNA repair. The effects of Jab1/CSN5's multiple protein interactions are generally oncogenic in nature, and overexpression of Jab1/CSN5 in cancer provides evidence that it is involved in the tumorigenic process. In this review, we highlight our current knowledge of Jab1/CSN5 function and the recent discoveries in dissecting the Jab1 signaling pathway. Further, we also discuss the regulation of Jab1/CSN5 in cancers and its potential as a therapeutic target.