EMAIL THIS PAGE TO A FRIEND
Drug metabolism reviews
In vivo and in vitro metabolic studies of furazolidone: a risk evaluation.
PMID 2102448
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9505
Furazolidone
C8H7N3O5
46297
Furazolidone, VETRANAL, analytical standard
C8H7N3O5