EMAIL THIS PAGE TO A FRIEND
AANA journal
Xenon as an anesthetic agent.
PMID 21067086
Abstract
Discovered in 1898 by British chemists, xenon is a rare gas belonging to the noble gases of the periodic table. Xenon is used in many different ways, from high-intensity lamps to jet propellant, and in 1939, its anesthetic properties were discovered. Xenon exerts its anesthetic properties, in part, through the noncompetitive inhibition of N-methyl-D-aspartate receptors. Currently, xenon is being used primarily throughout Europe; however, the high price of manufacturing and scavenging the noble gas has discouraged more widespread use. As technology in anesthetic delivery improves, xenon is being investigated further as a possible replacement for nitrous oxide as an inhalational agent. This article reviews the anesthetic properties of xenon and current and potential research about the gas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00472
Xenon, ≥99.995%
Xe