EMAIL THIS PAGE TO A FRIEND
Journal of photochemistry and photobiology. B, Biology
Transmembrane potential measurement with carbocyanine dye diS-C3-(5): fast fluorescence decay studies.
PMID 2107293
Abstract
The mechanism of carbocyanine dye diS-C3-(5) fluorescence intensity variations with transmembrane potential changes has been studied using time-resolved fluorescence spectroscopy. Clear evidence is given of the transmembrane-potential-dependent partition of the dye among various sites with different fluorescence lifetimes. It was found that fluorescence decay profiles reflect the transmembrane potential changes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43608
3,3′-Dipropylthiadicarbocyanine iodide, suitable for fluorescence, ≥98.0% (TLC)
C25H27IN2S2