EMAIL THIS PAGE TO A FRIEND
Angiogenesis
SR16388: a steroidal antiangiogenic agent with potent inhibitory effect on tumor growth in vivo.
PMID 21104121
Abstract
Angiogenesis is one of the major processes controlling growth and metastasis of tumors. Angiogenesis inhibitors have been targeted for the treatment of various cancers for more than 2 decades. We have developed a novel class of steroidal compounds aimed at blocking the angiogenic process in cancerous tissues. Our lead compound, SR16388, is a potent antiangiogenic agent with binding affinity to estrogen receptor-α (ER-α) and -β (ER-β) at the nanomolar range. This compound inhibited the proliferation of human microvascular endothelial cells (HMVEC) and various types of human cancer cells in vitro. SR16388 inhibited embryonic angiogenesis as measured in the chick chorioallantoic membrane (CAM) assay. The blood vessel density in the CAM was greatly reduced after the embryos were treated with 3xa0μg/CAM of SR16388 for 24xa0h. SR16388 at a dose of 2xa0μM prevented tube formation in Matrigel after HMVEC cells were treated for 8xa0h. In a modified Boyden chamber assay, SR16388 inhibited the migration of HMVECs by 80% at 500xa0nM. Using a novel in vivo Fibrin Z-chamber model, we demonstrated that SR16388 at a single daily oral dose of 3xa0mg/kg for 12 days significantly inhibited the granulation tissue (GT) thickness and the microvessel density of the GT as compared to control. More importantly, SR16388 down-regulated the pro-angiogenic transcription factors, hypoxia inducible factor 1α (HIF-1α) and signal transducer and activator of transcription 3 (STAT3) in non-small cell lung cancer (NSCLC) cells. Together, these effects of SR16388 can lead to the reduction of vascularization and tumor growth in vivo.
Related Materials