EMAIL THIS PAGE TO A FRIEND
Epilepsy research
Rufinamide for refractory epilepsy in a pediatric and young adult population.
PMID 21111576
Abstract
This chart review investigated the efficacy and safety of rufinamide in 45 children and young adults who experienced a broad spectrum of partial and generalized seizure/epilepsy types which have been refractory to therapy. Of these patients, 19 (46%) achieved a >50% decrease in seizure frequency on rufinamide, and 7 patients achieved a >75% decrease in seizure frequency. While 17 (37.8%) patients stopped their trial of rufinamide prior to the end of the review period, only 2 (4.4%) were due to adverse effects. Although additional research must be done, this data shows promise that rufinamide is a safe and efficacious adjunct for cases of refractory epilepsy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R8404
Rufinamide, ≥98% (HPLC), powder
C10H8F2N4O