EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Selective pure-phase synthesis of the multiferroic BaMF(4) (M = Mg, Mn, Co, Ni, and Zn) family.
PMID 21114317
Abstract
A low temperature, high yield hydrothermal route has been discovered for the phase-pure synthesis of the multiferroic BaMF(4) (M = Mg, Mn, Co, Ni, and Zn) family. The synthesis involves the use of CF(3)COOH instead of HF in an aqueous medium and, under the correct conditions, produces pure and polycrystalline BaMF(4). In addition to the synthetic description, second-harmonic generation, thermogravimetric, and differential scanning calorimetry data are presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
740969
Barium tetrafluorocobaltate, 99.9% trace metals basis
BaCoF4