EMAIL THIS PAGE TO A FRIEND
The international journal of tuberculosis and lung disease : the official journal of the International Union against Tuberculosis and Lung Disease
Methylene blue is a good background stain for tuberculosis light-emitting diode fluorescence microscopy.
PMID 21144242
Abstract
Damien Foundation Bangladesh tuberculosis (TB) control projects. To compare blue ink, potassium permanganate and methylene blue background staining for transmitted light-emitting diode (LED) TB fluorescence microscopy (FM). Auramine smears made in triplicate from Ziehl-Neelsen (ZN) acid-fast bacilli (AFB) positive or negative sputum and stained with one of the background variations were read blind by LED FM. Reference laboratory rechecking of discordant series was used before and after auramine restaining as the gold standard. Of 1977 series evaluated, 991 (50.1%) were made from ZN-positive specimens. There were 919, 942 and 958 FM true-positives with blue ink, permanganate and methylene blue counterstaining, against respectively 12, 12 and 16 false-positives. Methylene blue counterstaining was more sensitive (95.6%, 95%CI 94.2-96.8) than blue ink or permanganate (92.7%, 95%CI 90.9-94.3 and 93.6%, 95%CI 91.9-95.0; respectively P < 0.01 and < 0.05). No AFB could be found in 85% and 18% of 180 discordant series without and with restaining. Methylene blue is at least equivalent to potassium permanganate counterstaining for FM using blue LED transmitted excitation and is cheaper than blue ink. Restaining of all smears prior to first re-reading may be unavoidable for blinded rechecking of auramine-stained smears for external quality assessment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
66721
Methylene Blue zinc chloride double salt, for microscopy (Bact., Bot., Hist.), ~1 mol/mol water
C16H18ClN3S · 0.5ZnCl2 · xH2O