EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and antitumor activity of ureas containing pyrimidinyl group.
PMID 21144621
Abstract
A novel series of ureas containing pyrimidinyl group were designed and synthesized. Some of the prepared compounds showed potential cytotoxicity against several human cancer cell lines. From the structure-activity relationships we may conclude that introduction of a sulfide bridge between phenyl and pyrimidinyl rings would be critical for their biological activities.