EMAIL THIS PAGE TO A FRIEND
Biomaterials
Quantum dot-antisense oligonucleotide conjugates for multifunctional gene transfection, mRNA regulation, and tracking of biological processes.
PMID 21145105
Abstract
It was demonstrated that oligonucleotides, independent of their base sequence and length, could effectively induce the cellular uptake of mercapto acid-capped CdTe QDs after the oligonucleotides were covalently attached on the surface of the QDs. Following these experimental observations, a conjugate composed of covalently linked anti-survivin antisense oligonucleotides (ASON) and CdTe QDs was designed and synthesized. Then, the survivin mRNA down-regulation and the apoptosis of HeLa cells induced by ASON were studied. Systematic experimental results revealed that CdTe-ASON could effectively induce the apoptosis of HeLa cells, while CdTe QDs offered the possibility to visualize the specific intracellular localization of the CdTe-ASON probes strongly associated with their biological functions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8138
Chlorpromazine hydrochloride, ≥98% (TLC)
C17H19ClN2S · HCl