EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Enhanced efficacy of TD53, a novel algicidal agent, against the harmful algae via the liposomal delivery system.
PMID 21147206
Abstract
The present study aimed to design the liposomal delivery system for TD53, a novel algicial drug in order to improve the delivery properties of TD53 and evaluate its algicidal effects as well as selectivity against harmful and non-harmful algae. Liposomes of TD53 were prepared with 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) by a lyophilization, resulting in relatively small size vesicles (234±38nm) and narrow size distribution (PI=0.130±0.027). The drug leakage from the liposome was negligible in the F/2 media (<2% during 96h incubation). Subsequently algicidal activity of liposomal TD53 against harmful and nonharmful algae was evaluated at various concentrations. The IC(50) values of TD53 in liposome against harmful algae such as Chattonella marina, Heterosigma akashiwo and Cocholodinium polykrikoides were 2.675, 2.029, and 0.480μM, respectively, and were reduced by approximately 50% compared to those obtained from non-liposomal TD53. In contrast, the algicidal effect of liposomal TD53 was insignificant against non-harmful algae including Navicula pelliculosa, Nannochloropsis oculata and Phaeodactylum EPV. Those results suggested that liposomal delivery systems might be effective to enhance the efficacy of TD53 while maintaining the selectivity to harmful algal species.