EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and antibacterial evaluation of novel clarithromycin derivatives with C-4″ elongated arylalkyl groups against macrolide-resistant strains.
PMID 21159410
Abstract
Novel clarithromycin derivatives with C-4″ elongated arylalkyl groups were designed, synthesized and evaluated to probe the effect of different lengths of their C-4″ side chains on the activity against resistant bacterial strains. These derivatives had excellent activity against erythromycin-susceptible Streptococcus pneumoniae, Streptococcus aureus or Streptococcus pyogenes and some of them exhibited greatly improved activity against erythromycin-resistant strains. Compounds 18 and 16, which had the C-4″ elongated arylalkyl groups with eight atoms from the 4″-oxygen atom to the terminal benzene ring, were the most effective against S. pneumoniae expressing the erm gene and the erm and mef genes. In contrast, the most potent compounds 3, 5, 9, 17 and 18 against S. pneumoniae expressing the mef gene had C-4″ elongated arylalkyl groups with three to eight atoms between the 4″-oxygen atom and the terminal aromatic ring.