EMAIL THIS PAGE TO A FRIEND
Beilstein journal of organic chemistry
New library of aminosulfonyl-tagged Hoveyda-Grubbs type complexes: Synthesis, kinetic studies and activity in olefin metathesis transformations.
PMID 21165173
Abstract
Seven novel Hoveyda-Grubbs precatalysts bearing an aminosulfonyl function are reported. Kinetic studies indicate an activity enhancement compared to Hoveyda's precatalyst. A selection of these catalysts was investigated with various substrates in ring-closing metathesis of dienes or enynes and cross metathesis. The results demonstrate that these catalysts show a good tolerance to various chemical functions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
519367
3,5-Bis(trifluoromethyl)benzenesulfonyl chloride, 97%
C8H3ClF6O2S