EMAIL THIS PAGE TO A FRIEND
Virulence
Beyond ergosterol: linking pH to antifungal mechanisms.
PMID 21178501
Abstract
Many antifungal drugs including the highly successful azoles target the fungal-specific sterol, ergosterol, yet the molecular identity of cellular pathways mediating antifungal activity remained obscure. A recent study on the requirement of ergosterol in vacuolar H+-ATPase (V-ATPase) function uncovered a critical role for ion homeostasis downstream of azole inhibition of ergosterol biosynthesis. Here we review the functional link between regulation of V-ATPase by fungal membrane lipid components and discuss contributions of V-ATPase function to pathogenicity. The importance of pH homeostasis in virulence highlights its potential as target in antifungal chemotherapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1100000
Ergosterol, European Pharmacopoeia (EP) Reference Standard
C28H44O