EMAIL THIS PAGE TO A FRIEND
Biochemistry
Dithiothreitol causes HIV-1 integrase dimer dissociation while agents interacting with the integrase dimer interface promote dimer formation.
PMID 21222490
Abstract
We have developed a homogeneous time-resolved fluorescence resonance energy transfer (FRET)-based assay that detects the formation of HIV-1 integrase (IN) dimers. The assay utilizes IN monomers that express two different epitope tags that are recognized by their respective antibodies, coupled to distinct fluorophores. Surprisingly, we found that dithiothreitol (DTT), a reducing agent essential for in vitro enzymatic activity of IN, weakened the interaction between IN monomers. This effect of DTT on IN is dependent on its thiol groups, since the related chemical threitol, which contains hydroxyls in place of thiols, had no effect on IN dimer formation. By studying mutants of IN, we determined that cysteines in IN appear to be dispensable for the dimer dissociation effect of DTT. Peptides derived from the IN binding domain (IBD) of lens epithelium derived growth factor/transcriptional coactivator p75 (LEDGF), a cellular cofactor that interacts with the IN dimer interface, were tested in this IN dimerization assay. These peptides, which compete with LEDGF for binding to IN, displayed an intriguing equilibrium binding dose-response curve characterized by a plateau rising to a peak, then descending to a second plateau. Mathematical modeling of this binding system revealed that these LEDGF-derived peptides promote IN dimerization and block subunit exchange between IN dimers. This dose-response behavior was also observed with a small molecule that interacts with the IN dimer interface and inhibits LEDGF binding to IN. In conclusion, this novel IN dimerization assay revealed that peptide and small molecule inhibitors of the IN-LEDGF interaction also stabilize IN dimers and promote their formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E152
Epiboxidine hydrochloride
C10H14N2O · HCl