EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
ATR-mediated phosphorylation of DNA polymerase η is needed for efficient recovery from UV damage.
PMID 21242293
Abstract
DNA polymerase η (polη) belongs to the Y-family of DNA polymerases and facilitates translesion synthesis past UV damage. We show that, after UV irradiation, polη becomes phosphorylated at Ser601 by the ataxia-telangiectasia mutated and Rad3-related (ATR) kinase. DNA damage-induced phosphorylation of polη depends on its physical interaction with Rad18 but is independent of PCNA monoubiquitination. It requires the ubiquitin-binding domain of polη but not its PCNA-interacting motif. ATR-dependent phosphorylation of polη is necessary to restore normal survival and postreplication repair after ultraviolet irradiation in xeroderma pigmentosum variant fibroblasts, and is involved in the checkpoint response to UV damage. Taken together, our results provide evidence for a link between DNA damage-induced checkpoint activation and translesion synthesis in mammalian cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU013431 MISSION® esiRNA, esiRNA targeting human RAD18 (esiRNA1)