EMAIL THIS PAGE TO A FRIEND
Alcoholism, clinical and experimental research
Acetaldehyde burst protection of ADH1B*2 against alcoholism: an additional hormesis protection against esophageal cancers following alcohol consumption?
PMID 21284671
Abstract
This account of recent work presented at the 4th International Symposium on Alcohol Pancreatitis and Cirrhosis reports animal studies aimed at determining the role of the "acetaldehyde burst," generated shortly upon ethanol intake, as the mechanism of protection against alcoholism conferred by the ADH1B*2 polymorphism. Literature studies discussed suggest an additional role of the acetaldehyde burst on the paradoxical (hormesis) protection of the ADH1B*2 polymorphism against esophageal cancers in alcoholics.