EMAIL THIS PAGE TO A FRIEND
Physics in medicine and biology
Cone-beam optical computed tomography for gel dosimetry II: imaging protocols.
PMID 21285484
Abstract
This work develops imaging protocols for improved dose readout of a Fricke-xylenol orange-gelatin (FXG) gel-filled 1 L polyethylene terephthalate (PETE) jar dosimeter using a commercial Vista(TM) cone-beam optical computed tomography (CT) scanner from Modus Medical Devices Inc. (London, ON, Canada). To ensure good management of light source-detector stability, it was determined that (a) a minimum of 2 h warm-up time is necessary prior to dosimeter scanning, (b) the light source should be kept on until the completion of the last data scan except for the minimum amount of time required to acquire dark field images, and (c) the optional Vista software projection image normalization routine should be used in image reconstruction. The institution of dosimeter scan time and temperature control was strongly indicated from the experiments. A standard post-irradiation wait time of 30 min measured to within ±30 s was established to minimize the measurement uncertainties due to dosimeter development and diffusion. To alleviate thermochromic behavior leading to inaccurate dose readout, holding bath warm up and pre-scan temperature adjustment procedures were developed to control dosimeter temperature to within ±0.2 °C. The possibility of stray light minimizing protocols was also investigated and deemed to be unnecessary. The largest significant sources of stray light in the system were identified as being due to angled scatter from the dosimeter gelatin matrix and refraction from the jar wall interfaces. It was concluded that these phenomena would be better addressed through dosimeter modification and an inter-jar dose-to-attenuation calibration methodology, rather than by setting additional imaging protocols.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
258105
Sulfuric acid, ACS reagent, 95.0-98.0%
H2O4S