EMAIL THIS PAGE TO A FRIEND
Beilstein journal of organic chemistry
Hoveyda-Grubbs type metathesis catalyst immobilized on mesoporous molecular sieves MCM-41 and SBA-15.
PMID 21286390
Abstract
A commercially available Hoveyda-Grubbs type catalyst (RC303 Zhannan Pharma) was immobilized on mesoporous molecular sieves MCM-41 and on SBA-15 by direct interaction with the sieve wall surface. The immobilized catalysts exhibited high activity and nearly 100% selectivity in several types of alkene metathesis reactions. Ru leaching was found to depend on the substrate and solvent used (the lowest leaching was found for ring-closing metathesis of 1,7-octadiene in cyclohexane - 0.04% of catalyst Ru content). Results of XPS, UV-vis and NMR spectroscopy showed that at least 76% of the Ru content was bound to the support surface non-covalently and could be removed from the catalyst by washing with THF.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
474800
tert-Butyl N,N-diallylcarbamate, 98%
C11H19NO2