EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Design, synthesis and biological evaluation of novel 4-hydroxybenzene acrylic acid derivatives.
PMID 21288716
Abstract
A series of 4-hydroxybenzene acrylic acid derivatives were designed and synthesized based on the ferulic acid of natural active ingredients. The tested compound 5a, 5f and 6a have significant anti-inflammatory activity with suppression rates of 45.29%, 44.75% and 24.11%, respectively, compared with that of indomethacin, and their cardiac toxicity was not observed. The structure-function relationship shows that the p-hydroxyl group on the α-position benzene ring, particularly if acetylated, contributes to the considerable anti-inflammatory activity; that the carboxyl group on the double bond, if esterified, also contributes to the anti-inflammatory activity; that the p-methylsulfonyl group on the other benzene ring, whose introduction is due to the COX-2 selectivity, also contributes to anti-inflammatory activity surprisingly.