EMAIL THIS PAGE TO A FRIEND
Natural product communications
An efficient protocol for high-frequency direct multiple shoot regeneration from internodes of peppermint (Mentha x piperita).
PMID 21299127
Abstract
A simple, repeatable and efficient protocol for direct multiple shoot regeneration from internodal explants has been defined in peppermint (Mentha x piperita var. Indus). In vitro regenerated shoots of peppermint were excised into 4 to 8 mm long internodes and cultured on Murashige and Skoog's medium supplemented with different cytokinins. In the hormonal assay, 3.0 mg L(-1) zeatin or 6-isopentenyl adenine independently supplemented to half strength MS medium exhibited multiple shoot regeneration, while thiaduzorn (0.1-3.0 mg L(-1)) showed no morphogenetic effect. A maximum of 85% in vitro cultured explants showed multiple shoot formation with an average of 7 shoots per explant on MS medium supplemented with zeatin. Multiple shoots were initiated within three weeks of cultivation. Internodes with regenerated multiple shoots were transferred to half- strength MS medium without supplementing with any plant growth hormone for shoot elongation and rhizogenesis. Rooted plants acclimatized and grew to maturity under glasshouse conditions. The plantlets developed were phenotypically identical to the parent plant and exhibited 96% survival.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Z0164
Zeatin, BioReagent, plant cell culture tested, powder
C10H13N5O