EMAIL THIS PAGE TO A FRIEND
Natural product communications
Eugenol: a natural compound with versatile pharmacological actions.
PMID 21299140
Abstract
Eugenol, the major constituent of clove oil, has been widely used for its anesthetic and analgesic action in dentistry. Eugenol exhibits pharmacological effects on almost all systems and our aim is to review the research work that has identified these pharmacological actions. Eugenol possesses significant antioxidant, anti-inflammatory and cardiovascular properties, in addition to analgesic and local anesthetic activity. The metabolism and pharmacokinetics of the compound in humans have been studied. Eugenol has also been used as a penetration enhancer. The compound is a very promising candidate for versatile applications, and the design of new drugs based on the pharmacological effects of eugenol could be beneficial.