EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Design, synthesis, and biological evaluation of new cinnamic derivatives as antituberculosis agents.
PMID 21309577
Abstract
Tuberculosis, HIV coinfection with TB, emergence of multidrug-resistant TB, and extensively drug-resistant TB are the major causes of death from infectious diseases worldwide. Because no new drug has been introduced in the last several decades, new classes of molecules as anti-TB drugs are urgently needed. Herein, we report the synthesis and structure-activity relationships of a series of thioester, amide, hydrazide, and triazolophthalazine derivatives of 4-alkoxy cinnamic acid. Many compounds exhibited submicromolar minimum inhibitory concentrations against Mycobacterium tuberculosis strain (H(37)Rv). Interestingly, compound 13e, a 4-isopentenyloxycinnamyl triazolophthalazine derivative, was found to be 100-1800 times more active than isoniazid (INH) when tested for its ability to inhibit the growth of INH-resistant M. tuberculosis strains. The results also revealed that 13e does not interfere with mycolic acid biosynthesis, thereby pointing to a different mode of action and representing an attractive lead compound for the development of new anti-TB agents.