EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Microwave assisted one-pot synthesis of highly potent novel isoniazid analogues.
PMID 21320779
Abstract
A series of novel isoniazid (INH) analogues were synthesized by microwave assisted one pot reaction of INH, various benzaldehydes and dimedone in water with catalytic amount of DBSA. The synthesized compounds were evaluated for their anti-TB activity against Mycobacterium tuberculosis H37Rv (MTB) and multi-drug resistant Mycobacterium tuberculosis (MDR-TB). Among the 29 compounds, compound N-[9-[2-(benzyloxy)phenyl]-3,3,6,6-tetramethyl-1,8-dioxo-2,3,4,5,6,7,8,9-octahydro-10(1H)-acridinyl]isonicotinamide (12) inhibited MTB with MIC of <0.17 μM and MDR-TB with MIC of 0.69 μM.