EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Diels-Alder chemistry of graphite and graphene: graphene as diene and dienophile.
PMID 21341649
Abstract
The zero-band-gap electronic structure of graphene enables it to function as either the diene or the dienophile in the Diels-Alder reaction, and this versatile synthetic method offers a powerful strategy for the reversible modification of the electronic properties of graphene under very mild conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
276030
2,3-Dimethoxy-1,3-butadiene, 95%
C6H10O2