EMAIL THIS PAGE TO A FRIEND
Development (Cambridge, England)
Genetic mosaic analysis reveals a major role for frizzled 4 and frizzled 8 in controlling ureteric growth in the developing kidney.
PMID 21343368
Abstract
The developing mammalian kidney is an attractive system in which to study the control of organ growth. Targeted mutations in the Wnt receptors frizzled (Fz) 4 and Fz8 lead to reduced ureteric bud growth and a reduction in kidney size, a phenotype previously reported for loss of Wnt11. In cell culture, Fz4 and Fz8 can mediate noncanonical signaling stimulated by Wnt11, but only Fz4 mediates Wnt11-stimulated canonical signaling. In genetically mosaic mouse ureteric buds, competition between phenotypically mutant Fz4(-/-) or Fz4(-/-);Fz8(-/-) cells and adjacent phenotypically wild-type Fz4(+/-) or Fz4(+/-);Fz8(-/-) cells results in under-representation of the mutant cells to an extent far greater than would be predicted from the size reduction of homogeneously mutant kidneys. This discrepancy presumably reflects the compensatory action of a network of growth regulatory systems that minimize developmental perturbations. The present work represents the first description of a kidney phenotype referable to one or more Wnt receptors and demonstrates a general strategy for revealing the contribution of an individual growth regulatory pathway when it is part of a larger homeostatic network.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6278
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
C26H29NO2