EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Identification of novel indicators of cyclosporine A nephrotoxicity in a CD-1 mouse model.
PMID 21354196
Abstract
The calcineurin inhibitor cyclosporine A (CsA) is a widely used immunosuppressive agent. However, nephrotoxicity is a serious side effect observed in patients which limits clinical use of CsA. CsA nephrotoxicity is associated with tubulointerstitial injury progressing to nephropathy. This is typically diagnosed by invasive renal biopsy and is often only detected when the disease process is well advanced. Therefore identification of novel, early indicators of CsA nephrotoxicity could be clinically advantageous. This study aimed to establish a murine model of CsA nephrotoxicity and to identify urinary proteins that may indicate the onset of CsA-induced nephropathy using 2-D gel electrophoresis. CsA nephrotoxicity was induced in CD-1 mice by daily CsA administration for 4weeks. By week 4, elevated serum creatinine and proteinuria were observed after CsA treatment indicating significant renal dysfunction. Decreased cadherin-1, increased α-smooth muscle actin and fibroblast specific protein 1 in kidney tissue indicated disruption of normal tubular architecture. Alterations in podocin and uromodulin were also observed which may indicate damage to other segments of the nephron. Proteomic analysis of urine identified a number of differentially regulated proteins that may be involved in early CsA nephropathy including cadherin 1, superoxide dismutase and vinculin. These findings suggest novel mechanisms of CsA nephrotoxicity and identify novel potential markers of the disease.