EMAIL THIS PAGE TO A FRIEND
The Journal of experimental medicine
The human Tp53 Arg72Pro polymorphism explains different functional prognosis in stroke.
PMID 21357744
Abstract
The functional outcome after stroke is unpredictable; it is not accurately predicted by clinical pictures upon hospital admission. The presence of apoptotic neurons in the ischemic penumbra and perihematoma area may account for poor prognosis, but whether the highly variable stroke outcome reflects differences in genetic susceptibility to apoptosis is elusive. The p53 tumor suppressor protein, an important transcriptional regulator of apoptosis, naturally occurs in humans in two variants with single nucleotide polymorphisms resulting in Arg or Pro at residue 72. We show that poor functional outcome after either ischemic or hemorrhagic stroke was linked to the Arg/Arg genotype. This genotype was also associated with early neurological deterioration in ischemic stroke and with increased residual cavity volume in intracerebral hemorrhage. In primary cultured neurons, Arg(72)-p53, but not Pro(72)-p53, interacted directly with mitochondrial Bcl-xL and activated the intrinsic apoptotic pathway, increasing vulnerability to ischemia-induced apoptotic cell death. These results suggest that the Tp53 Arg/Arg genotype governs neuronal vulnerability to apoptosis and can be considered as a genetic marker predicting poor functional outcome after stroke.