PloS one
Prediction of Associations between microRNAs and Gene Expression in Glioma Biology.
PMID 21358821
Abstract
Despite progress in the determination of miR interactions, their regulatory role in cancer is only beginning to be unraveled. Utilizing gene expression data from 27 glioblastoma samples we found that the mere knowledge of physical interactions between specific mRNAs and miRs can be used to determine associated regulatory interactions, allowing us to identify 626 associated interactions, involving 128 miRs that putatively modulate the expression of 246 mRNAs. Experimentally determining the expression of miRs, we found an over-representation of over(under)-expressed miRs with various predicted mRNA target sequences. Such significantly associated miRs that putatively bind over-expressed genes strongly tend to have binding sites nearby the 3'UTR of the corresponding mRNAs, suggesting that the presence of the miRs near the translation stop site may be a factor in their regulatory ability. Our analysis predicted a significant association between miR-128 and the protein kinase WEE1, which we subsequently validated experimentally by showing that the over-expression of the naturally under-expressed miR-128 in glioma cells resulted in the inhibition of WEE1 in glioblastoma cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HUTR001C MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Housekeeping Control
HUTR002C
MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Empty Vector Control
HUTR003C MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Non-target Control 1
HUTR004C MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Non-target Control 2
HUTR00012 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, ARNTL
HUTR00018 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, RORA
HUTR00022 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, STAT3
HUTR00023 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TP53
HUTR00030 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TRIAP1
HUTR00036 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, MPP5
HUTR00041 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, ARFIP1
HUTR00043 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, DHX40
HUTR00046 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, LATS2
HUTR00047 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, RBL2
HUTR00238 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, ATP5G2
HUTR00247 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, NDST3
HUTR00256 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, WNK2
HUTR00264 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, EDF1
HUTR00276 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, XCR1
HUTR00286 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, RAB40A
HUTR00297 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TTR
HUTR00307 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, IFI27L1
HUTR00317 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, AMDHD2
HUTR00331 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, SHKBP1
HUTR00338 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, CCDC123
HUTR00345 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, CCT6B
HUTR00484 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TP73
HUTR00491 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TLR1
HUTR00499 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, SPATA5L1
HUTR00514 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, CGB1