EMAIL THIS PAGE TO A FRIEND
Reproduction in domestic animals = Zuchthygiene
The importance of adequate fixation for immunofluorescent staining of bovine embryos.
PMID 21366723
Abstract
Immunofluorescent staining is often used to investigate the expression of specific proteins in pre-implantation embryos. The success of this method is determined by the specificity of the antibodies, but also by the protocol used for fixation and permeabilization of the samples. In this study, different fixatives are compared in combination with immunofluorescent staining of caudal-type homeobox 2 (CDX2), fibronectin 1 (FN1) and integrins (ITGs) on bovine blastocysts. For both CDX2 and the ITGs, the outcome of the staining was largely dependent on the fixation methods. Paraformaldehyde fixation was best for the intracellular CDX2 protein, whereas acetone fixation gave the best results for the transmembrane ITGs. No difference was observed for the FN1 staining between samples fixed with paraformaldehyde or acetone. These examples demonstrate that the choice of fixation and permeabilization agents is very important for the outcome of the experiment, and this choice is dictated by the (extra)cellular location of the protein under investigation. Inappropriate fixation and/or permeabilization methods can lead to erroneous conclusions regarding the site and amount of protein expression.