EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
DNA-templated release of functional molecules with an azide-reduction-triggered immolative linker.
PMID 21369617
Abstract
Nucleic acid templated reactions have attracted significant attention for nucleic acid sensing. Herein we report a general design which extends the potential of nucleic acid templated reactions to unleash the function of a broad diversity of small molecules such as a transcription factor agonist, a cytotoxic or a fluorophore.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
191558
4-Aminobenzyl alcohol, 98%
C7H9NO