EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
A highly sensitive water-soluble system to sense glucose in aqueous solution.
PMID 21380440
Abstract
A glucose sensing switch is formed by water soluble conjugated polymer (PP-S-BINOL) and boronic acid-functionalized benzyl viologen (o-BBV). The two-component system shows a high sensitivity for glucose sensing with a 17-fold increase in the fluorescence intensity in the presence of 100 mM glucose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
104655
1,1′-Bi-2-naphthol, 99%
C20H14O2
679453
2-(Bromomethyl)phenylboronic acid
C7H8BBrO2