EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A flexible and stereoselective synthesis of azetidin-3-ones through gold-catalyzed intermolecular oxidation of alkynes.
PMID 21381165
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
758272
8-Ethylquinoline N-oxide, 97%
C11H11NO