EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Synthesis and biological evaluation of 3,7-diazabicyclo[4.3.0]nonan-8-ones as potential nootropic and analgesic drugs.
PMID 21381762
Abstract
A series of cis and trans 3,7-diazabicyclo[4.3.0]nonan-8-ones has been synthesized and tested for their ability to revert scopolamine-induced amnesia in the mouse passive-avoidance test. The racemates of the most potent compounds 4 and 7 were separated and tested, but no enantioselectivity was found for the nootropic activity. Compounds 4 and 7 and their enantiomers displayed interesting antihyperalgesic activity in two models of neuropathic pain (streptozotocin-induced and oxalilplatin-induced neuropathy) in comparison with pregabalin.
Related Materials