EMAIL THIS PAGE TO A FRIEND
Agents and actions
Role of Ca2(+)-ATPases in regulation of cellular Ca2+ signalling, as studied with the selective microsomal Ca2(+)-ATPase inhibitor, thapsigargin.
PMID 2139301