EMAIL THIS PAGE TO A FRIEND
Journal of proteomics
Identification of prognostic protein biomarkers in childhood acute lymphoblastic leukemia (ALL).
PMID 21396490
Abstract
Early response to 7 days of prednisolone (PRED) treatment is one of the important prognostic factors in predicting eventual outcome in childhood acute lymphoblastic leukemia (ALL). Using proteomic tools and clinically important leukemia cell lines (REH, 697, Sup-B15, RS4; 11), we have identified potential prognostic protein biomarkers as well as discovered promising regulators of PRED-induced apoptosis. After treatment with PRED, the four cell lines can be separated into resistant (REH) and sensitive (697, Sup-B15, RS4;11). Two dimensional gel electrophoresis (2-DE) and MALDI-TOF/TOF MS identified 77 and 17 significantly differentially expressed protein spots (p<0.05) in PRED-sensitive and PRED-resistant cell lines respectively. Several of these were validated by Western blot including proliferating cell nuclear antigen (PCNA), cofilin 1, voltage-dependent anion-channel protein 1 (VDAC1) and proteasome activator subunit 2 (PA28β). PCNA is a promising protein because of its important roles both in cell cycle regulation and survival control. We subsequently validated PCNA in 43 paired bone marrow samples from children with newly diagnosed ALL (Day 0) and 7 days after PRED treatment (Day 8). ROC curve analysis confirmed that PCNA was highly predictive of PRED response in patients (AUC=0.81, p=0.007) and most interestingly, independent of the molecular subtype, providing a promising universal prognostic marker.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8046
Mifepristone, ≥98%
C29H35NO2
P6004
Prednisolone, ≥99%
C21H28O5