EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Tumor necrosis factor up-regulates gamma-interferon binding in a human carcinoma cell line.
PMID 2141333
Abstract
WiDR colorectal carcinoma cells are highly sensitive to the synergistic cytotoxic effects of tumor necrosis factor (TNF) and gamma-interferon (IFN-gamma). In the present study, we have investigated the effects of recombinant human (rh) TNF and IFN-gamma on the binding of both ligands in this cell line. WiDR cells exhibited high affinity binding sites for both 125I-rhTNF (Kd = 1.66 x 10(-10) M, 920 sites/cell) and 125I-rhIFN-gamma (Kd = 4.15 x 10(-10) M, 18,960 sites/cell). Preincubation of the cells with rhTNF (24 h) increased cell-associated 125I-rhIFN-gamma radioactivity by 129% when binding was carried out at 37 degrees C, as a result of an increase in both surface bound and internalized 125I-rhIFN-gamma. However, rhTNF did not alter the degradation profile of released 125I-rhIFN-gamma radioactivity. Scatchard analysis of 125I-rhIFN-gamama binding data (4 degrees C) revealed that rhTNF induced a 245% increase in 125I-rhIFN-gamma binding sites. Conversely, rhIFN-gamma caused a 68% increase in 125I-rhTNF binding sites and a 58% increase in receptor affinity. rhIFN-gamma also increased the subsequent binding of 125I-rhIFN-gamma, whereas rhTNF increased the subsequent binding of 125I-rhTNF. Furthermore, preincubation of the cells with both rhTNF and rhIFN-gamma also resulted in an increase in the binding of both ligands. Actinomycin D and cycloheximide blocked all the effects of rhTNF and rhIFN-gamma on ligand binding. However, the basal level of 125I-rhIFN-gamma binding was insensitive to either inhibitor, whereas the basal level of 125I-rhTNF binding was decreased by both inhibitors. These data indicate that in some cell types TNF and IFN-gamma may induce an increase in their own receptors (homologous up-regulation) and concomitantly increase each other's receptors (heterologous up-regulation) and that these actions are due, in part, to enhanced receptor synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85111501
WiDr