EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Myospryn is a calcineurin-interacting protein that negatively modulates slow-fiber-type transformation and skeletal muscle regeneration.
PMID 21427212
Abstract
The calcium-calmodulin-regulated protein phosphatase calcineurin plays an important regulatory role in muscle differentiation, fiber-type determination, hypertrophy, and muscle regeneration. Because calcineurin functions in numerous processes in muscle, multiple mechanisms are likely necessary to ensure that the activity of this phosphatase is appropriately regulated. Here we demonstrate that the muscle-specific scaffolding protein myospryn modulates calcineurin signaling by inhibiting calcineurin-dependent transcriptional activity in C2C12 myoblasts through direct interaction with the enzyme via its noncanonical tripartite motif (TRIM-like). Consistent with these data, transgenic mice overexpressing both the TRIM-like domain of myospryn and constitutively active calcineurin displayed a severe attenuation in the ability of calcineurin to induce a slow-fiber phenotype. Furthermore, transgenic mice overexpressing the TRIM-like domain of myospryn displayed attenuated muscle regeneration after cardiotoxin-induced muscle injury. These results indicate that myospryn functions as a novel inhibitor of the calcineurin signaling pathway in skeletal muscle.
Related Materials