EMAIL THIS PAGE TO A FRIEND
American journal of hypertension
A68930: a potent and specific agonist for the D-1 dopamine receptor.
PMID 2143387
Abstract
A68930 (5,6-dihydroxy-3-phenyl-1-aminomethyl-isochroman) is a potent (EC50 = 2.5 nmol/L) partial agonist at the D-1 dopamine receptor. In contrast, A68930 is a much weaker agonist (EC50 = 3,920 nmol/L) at the D-2 dopamine receptor. The orientation of the 3-phenyl substituent in the molecule is critical for the affinity and selectivity of the molecule towards the D-1 receptor in vitro and in vivo. The role of the D-1 receptor in the functioning of the basal ganglia is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8852
A-68930 hydrochloride, ≥98% (HPLC), powder
C16H17NO3 · HCl