EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Amberlite-IRA-402 (OH) ion exchange resin mediated synthesis of indolizines, pyrrolo [1,2-a] quinolines and isoquinolines: antibacterial and antifungal evaluation of the products.
PMID 21440339
Abstract
A number of indolizines and pyrrolo[1,2-a]quinolines/isoquinolines were prepared from phenacyl pyridinium, quinolinium and isoquinolinium salts derived from the reaction of the heterocycles with 2-bromo acetophenone with alkynes and alkenes using amberlite-IRA-402 (OH) ion exchange resin as the base. Antibacterial and antifungal studies were carried out against thirteen bacterial and four fungal strains, which revealed that three derivatives (4a, 4b, 7a) out of fifteen are effective against all the thirteen strains and one derivative, 10, showed dual antibactericidal and antifungal efficacy.