EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A broadly applicable copper reagent for trifluoromethylations and perfluoroalkylations of aryl iodides and bromides.
PMID 21442711