EMAIL THIS PAGE TO A FRIEND
Journal of clinical immunology
Distribution of CD45RA and CD45RO T-lymphocyte subsets in rheumatoid arthritis synovial tissue.
PMID 2145318
Abstract
We have characterized the lymphocytes in the synovium of patients with rheumatoid arthritis (RA) by immunohistochemistry using monoclonal antibodies directed against B lymphocytes, T lymphocytes, and antibodies directed against CD45RA and CD45RO, which define T-cell subsets. Both CD45RA+ and CD45RO+ T lymphocytes were detected in the perivascular regions. CD45RA+ lymphocytes were present primarily in perivascular areas of moderate to large lymphocytic infiltration. Some synovial perivascular lymphocytic aggregates were organized into focal areas of CD45RA+ B lymphocytes surrounded by CD45RO+ T lymphocytes. In areas of diffuse lymphocytic infiltration, the T lymphocytes were CD45RO+. These data suggest that both CD45RO+ and CD45RA+ T lymphocytes enter the RA synovial tissue via the synovial vasculature and that, once in the tissue, the CD45RA+ T lymphocytes may undergo activation/maturation and acquire the CD45RO phenotype.